The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031421.3 | NP_113609.1 | 6 - 7 | 957 | 82 |
XM_011525323.2 | XP_011523625.1 | 6 - 7 | 897 | 82 | |
XM_011525324.2 | XP_011523626.1 | 6 - 7 | 924 | 82 | |
GenBank mRNA | AK055498.1 | - | 6 - 7 | 902 | 82 |
AL136760.1 | - | 6 - 7 | 924 | 82 | |
BC067297.1 | - | 6 - 7 | 916 | 82 |