Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VNN1
Entrez Gene ID
8876
Gene Name
vanin 1
Gene Aliases
HDLCQ8, Tiff66
Chromosome Location
Chr.6: 132680858 - 132714055 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004666.2 NP_004657.2 5 - 6 1209 82
GenBank mRNA AJ132099.1 - 5 - 6 1203 82
AK290425.1 - 5 - 6 1206 82
AK314265.1 - 5 - 6 1206 82
BC096265.3 - 5 - 6 1203 82
BC096266.3 - 5 - 6 1203 82
BC096267.3 - 5 - 6 1203 82
BC096268.1 - 5 - 6 1202 82
U39664.1 - 5 - 6 1204 82

View Full Product Details