The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001226.3 | NP_001217.2 | 5 - 6 | 564 | 82 |
NM_032992.2 | NP_116787.1 | 2 - 3 | 297 | 82 | |
NR_133012.1 | 4 - 5 | 487 | 82 | ||
GenBank mRNA | BC000305.2 | - | 5 - 6 | 512 | 82 |
BC004460.1 | - | 5 - 6 | 512 | 82 | |
BM837197.1 | - | 2 - 3 | 243 | 82 | |
BX380795.2 | - | 4 - 5 | 428 | 82 |