The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024674.2 | NP_001019845.1 | 1 - 2 | 42 | 83 |
XM_011537320.2 | XP_011535622.1 | 1 - 2 | 164 | 83 | |
XM_011537321.2 | XP_011535623.1 | 1 - 2 | 128 | 83 | |
XM_011537322.2 | XP_011535624.2 | 1 - 2 | 124 | 83 | |
XM_017021764.1 | XP_016877253.1 | 1 - 2 | 103 | 83 | |
GenBank mRNA | AK023979.1 | - | 1 - 2 | 30 | 83 |
AK307062.1 | - | 1 - 2 | 39 | 83 | |
BC094003.1 | - | 1 - 2 | 199 | 83 | |
BC094005.1 | - | 1 - 2 | 199 | 83 |