Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLL2
Entrez Gene ID
7093
Gene Name
tolloid like 2
Gene Aliases
-
Chromosome Location
Chr.10: 96364606 - 96513926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012465.3 NP_036597.1 12 - 13 1772 75
GenBank mRNA AB023149.2 - 12 - 13 1722 75
AF059516.1 - 12 - 13 1895 75
AK026106.1 - 12 - 13 1698 75
BC013871.2 - 12 - 13 1732 75
BC112341.1 - 12 - 13 1655 75
BC112366.1 - 12 - 13 1655 75
BC113577.1 - 12 - 13 1588 75

View Full Product Details