The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293274.1 | NP_001280203.1 | 19 - 20 | 3118 | 67 |
NM_020925.2 | NP_065976.2 | 20 - 21 | 2839 | 67 | |
XM_011541862.1 | XP_011540164.1 | 19 - 20 | 3114 | 67 | |
XM_011541863.1 | XP_011540165.1 | 20 - 21 | 3263 | 67 | |
GenBank mRNA | AB046793.1 | - | 18 - 19 | 2621 | 67 |
AK022571.1 | - | 7 - 8 | 937 | 67 | |
AK024256.1 | - | 10 - 11 | 1264 | 67 | |
AK299361.1 | - | 6 - 7 | 682 | 67 | |
AL833000.1 | - | 10 - 11 | 1288 | 67 | |
BC039301.1 | - | 20 - 21 | 2825 | 67 | |
BX537603.1 | - | 19 - 20 | 3117 | 67 |