Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 64470793 - 64693058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293274.1 NP_001280203.1 24 - 25 3718 92
NM_020925.2 NP_065976.2 25 - 26 3439 92
XM_011541862.1 XP_011540164.1 24 - 25 3714 92
XM_011541863.1 XP_011540165.1 25 - 26 3863 92
GenBank mRNA AB046793.1 - 23 - 24 3221 92
AK022571.1 - 12 - 13 1537 92
AK024256.1 - 15 - 16 1864 92
AL833000.1 - 15 - 16 1888 92
BC039301.1 - 25 - 26 3425 92
BX537603.1 - 24 - 25 3717 92

View Full Product Details