Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location
Chr.11: 47465937 - 47565569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025596.2 NP_001020767.1 11 - 12 1339 107
NM_001172639.1 NP_001166110.1 14 - 15 2301 107
NM_001172640.1 NP_001166111.1 11 - 12 1336 107
NM_006560.3 NP_006551.1 12 - 13 1576 107
NM_198700.2 NP_941989.1 12 - 13 1521 107
XM_011519848.2 XP_011518150.1 12 - 13 1414 107
XM_011519850.2 XP_011518152.2 12 - 13 1650 107
XM_011519852.2 XP_011518154.1 14 - 15 1762 107
XM_011519854.1 XP_011518156.1 16 - 17 1870 107
XM_011519855.1 XP_011518157.1 12 - 13 1591 107
XM_011519856.1 XP_011518158.1 13 - 14 1672 107
XM_011519857.1 XP_011518159.1 12 - 13 1543 107
XM_011519858.1 XP_011518160.1 12 - 13 1638 107
XM_011519859.1 XP_011518161.1 16 - 17 1858 107
XM_017017101.1 XP_016872590.1 15 - 16 1754 107
XM_017017102.1 XP_016872591.1 14 - 15 1673 107
XM_017017103.1 XP_016872592.1 13 - 14 1566 107
XM_017017104.1 XP_016872593.1 13 - 14 1566 107
XM_017017105.1 XP_016872594.1 13 - 14 1563 107
XM_017017106.1 XP_016872595.1 15 - 16 1751 107
XM_017017107.1 XP_016872596.1 13 - 14 1563 107
XM_017017108.1 XP_016872597.1 13 - 14 1554 107
XM_017017109.1 XP_016872598.1 12 - 13 1641 107
XM_017017110.1 XP_016872599.1 13 - 14 1554 107
XM_017017111.1 XP_016872600.1 13 - 14 1551 107
XM_017017112.1 XP_016872601.1 13 - 14 1551 107
XM_017017113.1 XP_016872602.1 13 - 14 1551 107
XM_017017114.1 XP_016872603.1 13 - 14 1551 107
XM_017017115.1 XP_016872604.1 14 - 15 1594 107
XM_017017116.1 XP_016872605.1 13 - 14 1629 107
XM_017017117.1 XP_016872606.1 12 - 13 1390 107
XM_017017118.1 XP_016872607.1 12 - 13 1512 107
XM_017017119.1 XP_016872608.1 14 - 15 1731 107
XM_017017120.1 XP_016872609.1 12 - 13 1543 107
XM_017017121.1 XP_016872610.1 12 - 13 1638 107
XM_017017122.1 XP_016872611.1 12 - 13 1543 107
XM_017017123.1 XP_016872612.1 12 - 13 1591 107
XM_017017124.1 XP_016872613.1 14 - 15 1682 107
XM_017017125.1 XP_016872614.1 12 - 13 1411 107
XM_017017126.1 XP_016872615.1 12 - 13 1540 107
XM_017017127.1 XP_016872616.1 12 - 13 1564 107
XM_017017128.1 XP_016872617.1 12 - 13 1588 107
XM_017017129.1 XP_016872618.1 13 - 14 1586 107
XM_017017130.1 XP_016872619.1 12 - 13 1540 107
XM_017017131.1 XP_016872620.1 12 - 13 1564 107
XM_017017132.1 XP_016872621.1 12 - 13 1588 107
XM_017017133.1 XP_016872622.1 12 - 13 1370 107
XM_017017134.1 XP_016872623.1 13 - 14 1509 107
XM_017017135.1 XP_016872624.1 12 - 13 1528 107
GenBank mRNA AB210019.1 - 14 - 15 2301 107
AF248648.1 - 11 - 12 1347 107
AF267533.1 - 11 - 12 1340 107
AF267534.1 - 11 - 12 1331 107
AJ007988.1 - 11 - 12 1329 107
AK304430.1 - 11 - 12 1279 107
AK307679.1 - 6 - 7 801 107
BC031079.1 - 12 - 13 1521 107
U63289.1 - 12 - 13 1454 107

View Full Product Details