The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011519854.1 | XP_011518156.1 | 4 - 5 | 417 | 115 |
XM_011519859.1 | XP_011518161.1 | 4 - 5 | 417 | 115 | |
XM_017017124.1 | XP_016872613.1 | 2 - 3 | 232 | 115 |