Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FNBP4
Entrez Gene ID
23360
Gene Name
formin binding protein 4
Gene Aliases
FBP30
Chromosome Location
Chr.11: 47716510 - 47767478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318339.1 NP_001305268.1 13 - 14 2513 119
NM_015308.3 NP_056123.2 13 - 14 2507 119
GenBank mRNA AB023231.2 - 13 - 14 2506 119
AK022987.1 - 6 - 7 951 119
AK054636.1 - 13 - 14 2330 119
AK095093.1 - 1 - 2 2301 119
AK123898.1 - 13 - 14 2271 119
AL137480.1 - 13 - 14 2306 119
BC037404.1 - 13 - 14 2338 119
BX537864.1 - 1 - 2 2430 119

View Full Product Details