The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318339.1 | NP_001305268.1 | 6 - 7 | 1103 | 85 |
NM_015308.3 | NP_056123.2 | 6 - 7 | 1097 | 85 | |
GenBank mRNA | AB023231.2 | - | 6 - 7 | 1096 | 85 |
AK054636.1 | - | 6 - 7 | 920 | 85 | |
AK123898.1 | - | 5 - 6 | 764 | 85 | |
AL137480.1 | - | 6 - 7 | 896 | 85 | |
BC037404.1 | - | 6 - 7 | 928 | 85 |