The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081686.1 | NP_001075155.1 | 2 - 3 | 422 | 71 |
NM_021176.2 | NP_066999.1 | 2 - 3 | 422 | 71 | |
GenBank mRNA | BC104778.1 | - | 2 - 3 | 422 | 71 |
BC113376.1 | - | 2 - 3 | 422 | 71 | |
BQ268961.1 | - | 2 - 3 | 357 | 71 |