Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
Chromosome Location
Chr.12: 121402289 - 121582268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005366.1 NP_001005366.1 13 - 14 2272 90
NM_032590.4 NP_115979.3 13 - 14 2062 90
XM_005253955.3 XP_005254012.1 13 - 14 2272 90
XM_005253956.3 XP_005254013.1 13 - 14 1869 90
XM_005253961.4 XP_005254018.1 2 - 3 357 90
XM_011538867.2 XP_011537169.1 13 - 14 3315 90
XM_011538868.2 XP_011537170.1 13 - 14 3311 90
XM_011538869.2 XP_011537171.1 13 - 14 3313 90
XM_011538875.2 XP_011537177.1 2 - 3 357 90
GenBank mRNA AB031230.1 - 3 - 4 521 90
AJ459424.1 - 13 - 14 2034 90
AK027440.1 - 3 - 4 451 90
AK127328.1 - 13 - 14 2272 90
AK299827.1 - 2 - 3 367 90
AK316470.1 - 2 - 3 369 90
AL133572.1 - 9 - 10 1575 90
BC115380.1 - 13 - 14 2001 90

View Full Product Details