Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPCN2
Entrez Gene ID
219931
Gene Name
two pore segment channel 2
Gene Aliases
SHEP10, TPC2
Chromosome Location
Chr.11: 69048876 - 69090604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139075.3 NP_620714.2 14 - 15 1465 68
XM_005273824.3 XP_005273881.1 14 - 15 1447 68
XM_005273826.3 XP_005273883.1 12 - 13 1158 68
XM_005273830.3 XP_005273887.1 13 - 14 1289 68
XM_005273831.3 XP_005273888.1 11 - 12 1141 68
XM_005273832.3 XP_005273889.1 13 - 14 1366 68
XM_011544802.2 XP_011543104.1 13 - 14 1338 68
XM_011544805.1 XP_011543107.1 14 - 15 1450 68
XM_011544806.1 XP_011543108.1 14 - 15 1450 68
XM_011544807.2 XP_011543109.1 13 - 14 1286 68
XM_011544808.2 XP_011543110.1 11 - 12 1216 68
XM_017017328.1 XP_016872817.1 12 - 13 1281 68
XM_017017329.1 XP_016872818.1 12 - 13 1278 68
XM_017017330.1 XP_016872819.1 13 - 14 1519 68
XM_017017331.1 XP_016872820.1 11 - 12 1689 68
XM_017017332.1 XP_016872821.1 13 - 14 1465 68
XM_017017333.1 XP_016872822.1 11 - 12 1199 68
XM_017017334.1 XP_016872823.1 9 - 10 944 68
XM_017017335.1 XP_016872824.1 8 - 9 879 68
XM_017017336.1 XP_016872825.1 7 - 8 805 68
GenBank mRNA AB209253.1 - 9 - 10 942 68
AK023366.1 - 13 - 14 1344 68
AK123089.1 - 12 - 13 1158 68
AK298919.1 - 14 - 15 1386 68
AL137479.1 - 2 - 3 208 68
AY029200.1 - 14 - 15 1450 68
BC063008.1 - 14 - 15 1401 68

View Full Product Details