Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPCN2
Entrez Gene ID
219931
Gene Name
two pore segment channel 2
Gene Aliases
SHEP10, TPC2
Chromosome Location
Chr.11: 69048876 - 69090604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139075.3 NP_620714.2 4 - 5 547 87
XM_005273824.3 XP_005273881.1 4 - 5 532 87
XM_005273830.3 XP_005273887.1 4 - 5 444 87
XM_005273832.3 XP_005273889.1 4 - 5 551 87
XM_011544802.2 XP_011543104.1 3 - 4 420 87
XM_011544805.1 XP_011543107.1 4 - 5 532 87
XM_011544806.1 XP_011543108.1 4 - 5 532 87
XM_011544807.2 XP_011543109.1 4 - 5 444 87
XM_011544808.2 XP_011543110.1 3 - 4 474 87
XM_017017328.1 XP_016872817.1 4 - 5 532 87
XM_017017329.1 XP_016872818.1 4 - 5 532 87
XM_017017330.1 XP_016872819.1 4 - 5 704 87
XM_017017331.1 XP_016872820.1 2 - 3 874 87
XM_017017332.1 XP_016872821.1 4 - 5 550 87
XM_017017333.1 XP_016872822.1 4 - 5 553 87
GenBank mRNA AB209253.1 - 1 - 2 193 87
AK023366.1 - 4 - 5 532 87
AK298919.1 - 4 - 5 468 87
AY029200.1 - 4 - 5 532 87
BC063008.1 - 4 - 5 483 87

View Full Product Details