Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBNL1
Entrez Gene ID
4154
Gene Name
muscleblind like splicing regulator 1
Gene Aliases
EXP, MBNL
Chromosome Location
Chr.3: 152243656 - 152465780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314057.1 NP_001300986.1 5 - 6 1174 65
NM_021038.4 NP_066368.2 4 - 5 1959 65
NM_207292.2 NP_997175.1 4 - 5 1959 65
NM_207293.1 NP_997176.1 3 - 4 1334 65
NM_207296.1 NP_997179.1 3 - 4 1334 65
NM_207297.1 NP_997180.1 3 - 4 1334 65
XM_005247457.4 XP_005247514.1 5 - 6 1527 65
XM_005247458.4 XP_005247515.1 5 - 6 2781 65
XM_005247460.4 XP_005247517.1 5 - 6 1490 65
XM_005247461.4 XP_005247518.1 5 - 6 1527 65
XM_005247462.4 XP_005247519.1 5 - 6 1527 65
XM_005247463.4 XP_005247520.1 5 - 6 1527 65
XM_005247464.4 XP_005247521.1 4 - 5 1585 65
XM_005247465.4 XP_005247522.1 5 - 6 1527 65
XM_005247466.4 XP_005247523.1 4 - 5 1562 65
XM_005247467.4 XP_005247524.1 5 - 6 1527 65
XM_005247468.4 XP_005247525.1 5 - 6 1527 65
XM_005247471.4 XP_005247528.1 4 - 5 1557 65
XM_005247474.3 XP_005247531.1 3 - 4 445 65
XM_005247475.3 XP_005247532.1 4 - 5 768 65
XM_005247476.4 XP_005247533.1 3 - 4 535 65
XM_005247477.4 XP_005247534.1 3 - 4 534 65
XM_006713639.1 XP_006713702.1 5 - 6 1509 65
XM_011512848.2 XP_011511150.1 5 - 6 1131 65
XM_017006423.1 XP_016861912.1 4 - 5 1778 65
XM_017006424.1 XP_016861913.1 4 - 5 1605 65
XM_017006425.1 XP_016861914.1 4 - 5 1607 65
XM_017006426.1 XP_016861915.1 4 - 5 1605 65
XM_017006427.1 XP_016861916.1 4 - 5 1752 65
XM_017006428.1 XP_016861917.1 4 - 5 1416 65
XM_017006429.1 XP_016861918.1 4 - 5 1499 65
XM_017006430.1 XP_016861919.1 4 - 5 1563 65
XM_017006431.1 XP_016861920.1 4 - 5 1640 65
XM_017006432.1 XP_016861921.1 4 - 5 1562 65
XM_017006433.1 XP_016861922.1 4 - 5 1562 65
XM_017006434.1 XP_016861923.1 4 - 5 1568 65
XM_017006435.1 XP_016861924.1 4 - 5 575 65
XM_017006436.1 XP_016861925.1 4 - 5 2688 65
XM_017006437.1 XP_016861926.1 4 - 5 1743 65
XM_017006439.1 XP_016861928.1 4 - 5 768 65
XM_017006440.1 XP_016861929.1 3 - 4 502 65
XM_017006441.1 XP_016861930.1 3 - 4 535 65
XM_017006442.1 XP_016861931.1 3 - 4 638 65
XM_017006443.1 XP_016861932.1 3 - 4 646 65
GenBank mRNA AB007888.1 - 4 - 5 1959 65
AF401998.1 - 3 - 4 545 65
AF497718.1 - 3 - 4 545 65
AF497719.1 - 3 - 4 545 65
AJ308400.1 - 3 - 4 574 65
AK304089.1 - 2 - 3 254 65
AK307420.1 - 2 - 3 254 65
BC043493.1 - 4 - 5 1464 65
BC050535.1 - 5 - 6 695 65
Y13829.1 - 3 - 4 1056 65

View Full Product Details