The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000693.3 | NP_000684.2 | 5 - 6 | 758 | 71 |
GenBank mRNA | AK312303.1 | - | 5 - 6 | 619 | 71 |
BC069274.1 | - | 5 - 6 | 598 | 71 | |
U07919.1 | - | 5 - 6 | 594 | 71 |