Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALDH1A3
Entrez Gene ID
220
Gene Name
aldehyde dehydrogenase 1 family member A3
Gene Aliases
ALDH1A6, ALDH6, MCOP8, RALDH3
Chromosome Location
Chr.15: 100879692 - 100916626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000693.3 NP_000684.2 8 - 9 1100 64
NM_001293815.1 NP_001280744.1 5 - 6 779 64
GenBank mRNA AK302607.1 - 7 - 8 721 64
AK312303.1 - 8 - 9 961 64
BC069274.1 - 8 - 9 940 64
BX538027.1 - 5 - 6 597 64
U07919.1 - 8 - 9 936 64

View Full Product Details