Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS7
Entrez Gene ID
6000
Gene Name
regulator of G-protein signaling 7
Gene Aliases
-
Chromosome Location
Chr.1: 240775193 - 241357374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282773.1 NP_001269702.1 8 - 9 914 130
NM_001282775.1 NP_001269704.1 10 - 11 1073 130
NM_001282778.1 NP_001269707.1 10 - 11 1073 130
NM_002924.5 NP_002915.3 10 - 11 1073 130
XM_005273218.3 XP_005273275.1 10 - 11 1217 130
XM_006711800.3 XP_006711863.1 9 - 10 1008 130
XM_011544246.2 XP_011542548.1 9 - 10 667 130
XM_011544247.2 XP_011542549.1 9 - 10 712 130
XM_017002001.1 XP_016857490.1 9 - 10 1166 130
XM_017002002.1 XP_016857491.1 11 - 12 1092 130
XM_017002003.1 XP_016857492.1 9 - 10 1166 130
XM_017002004.1 XP_016857493.1 9 - 10 1166 130
XM_017002005.1 XP_016857494.1 10 - 11 2108 130
XM_017002006.1 XP_016857495.1 11 - 12 1138 130
XM_017002007.1 XP_016857496.1 11 - 12 1069 130
XM_017002008.1 XP_016857497.1 11 - 12 1097 130
XM_017002009.1 XP_016857498.1 9 - 10 997 130
XM_017002010.1 XP_016857499.1 10 - 11 782 130
XM_017002011.1 XP_016857500.1 7 - 8 2179 130
XM_017002012.1 XP_016857501.1 7 - 8 2177 130
XM_017002013.1 XP_016857502.1 7 - 8 2176 130
GenBank mRNA AF090116.1 - 10 - 11 928 130
AF090117.1 - 3 - 4 184 130
AF493930.1 - 7 - 8 130
AF493931.1 - 9 - 10 691 130
AK294234.1 - 10 - 11 907 130
AK294354.1 - 10 - 11 1048 130
AK314862.1 - 10 - 11 934 130
AY587875.1 - 9 - 10 691 130
BC022009.1 - 10 - 11 1026 130
U32439.1 - 8 - 9 545 130

View Full Product Details