The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282773.1 | NP_001269702.1 | 13 - 14 | 1499 | 103 |
NM_001282775.1 | NP_001269704.1 | 15 - 16 | 1658 | 103 | |
NM_001282778.1 | NP_001269707.1 | 15 - 16 | 1658 | 103 | |
NM_002924.5 | NP_002915.3 | 15 - 16 | 1658 | 103 | |
XM_005273218.3 | XP_005273275.1 | 15 - 16 | 1802 | 103 | |
XM_006711800.3 | XP_006711863.1 | 14 - 15 | 1593 | 103 | |
XM_011544246.2 | XP_011542548.1 | 14 - 15 | 1252 | 103 | |
XM_011544247.2 | XP_011542549.1 | 14 - 15 | 1297 | 103 | |
XM_017002001.1 | XP_016857490.1 | 14 - 15 | 1751 | 103 | |
XM_017002002.1 | XP_016857491.1 | 16 - 17 | 1677 | 103 | |
XM_017002003.1 | XP_016857492.1 | 14 - 15 | 1751 | 103 | |
XM_017002004.1 | XP_016857493.1 | 14 - 15 | 1751 | 103 | |
XM_017002005.1 | XP_016857494.1 | 15 - 16 | 2693 | 103 | |
XM_017002006.1 | XP_016857495.1 | 16 - 17 | 1723 | 103 | |
XM_017002007.1 | XP_016857496.1 | 16 - 17 | 1654 | 103 | |
XM_017002008.1 | XP_016857497.1 | 16 - 17 | 1682 | 103 | |
XM_017002009.1 | XP_016857498.1 | 14 - 15 | 1582 | 103 | |
XM_017002010.1 | XP_016857499.1 | 15 - 16 | 1367 | 103 | |
XM_017002011.1 | XP_016857500.1 | 12 - 13 | 2764 | 103 | |
XM_017002012.1 | XP_016857501.1 | 12 - 13 | 2762 | 103 | |
XM_017002013.1 | XP_016857502.1 | 12 - 13 | 2761 | 103 | |
GenBank mRNA | AF090116.1 | - | 15 - 16 | 1513 | 103 |
AF090117.1 | - | 8 - 9 | 769 | 103 | |
AF493930.1 | - | 12 - 13 | 1117 | 103 | |
AF493931.1 | - | 14 - 15 | 1276 | 103 | |
AK294234.1 | - | 15 - 16 | 1492 | 103 | |
AK294354.1 | - | 15 - 16 | 1633 | 103 | |
AK314862.1 | - | 15 - 16 | 1519 | 103 | |
AY587875.1 | - | 14 - 15 | 1276 | 103 | |
BC022009.1 | - | 15 - 16 | 1611 | 103 | |
U32439.1 | - | 13 - 14 | 1130 | 103 |