Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
Chromosome Location
Chr.11: 35431827 - 35530300 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001991.2 NP_001001991.1 1 - 2 515 71
NM_001282676.1 NP_001269605.1 1 - 2 515 71
NM_015430.3 NP_056245.2 1 - 2 515 71
GenBank mRNA AK027841.1 - 1 - 2 115 71
AK300185.1 - 1 - 2 100 71
AK316188.1 - 1 - 2 87 71
AL832391.1 - 1 - 2 81 71
AY358346.1 - 1 - 2 94 71
BC089434.1 - 1 - 2 107 71
BX640676.1 - 1 - 2 515 71

View Full Product Details