The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000328.2 | NP_000319.1 | 5 - 6 | 638 | 76 |
NM_001034853.1 | NP_001030025.1 | 5 - 6 | 638 | 76 | |
XM_005272633.2 | XP_005272690.1 | 5 - 6 | 667 | 76 | |
XM_011543940.2 | XP_011542242.1 | 5 - 6 | 637 | 76 | |
XM_017029710.1 | XP_016885199.1 | 5 - 6 | 667 | 76 | |
XM_017029711.1 | XP_016885200.1 | 5 - 6 | 667 | 76 | |
XM_017029712.1 | XP_016885201.1 | 5 - 6 | 667 | 76 | |
GenBank mRNA | AJ238395.1 | - | 5 - 6 | 525 | 76 |
AK223491.1 | - | 5 - 6 | 612 | 76 | |
AK291832.1 | - | 5 - 6 | 645 | 76 | |
BC031624.1 | - | 5 - 6 | 609 | 76 | |
BX644678.1 | - | 5 - 6 | 637 | 76 | |
KU178311.1 | - | 5 - 6 | 470 | 76 | |
KU178312.1 | - | 5 - 6 | 470 | 76 | |
U57629.1 | - | 5 - 6 | 529 | 76 | |
X97668.1 | - | 5 - 6 | 470 | 76 |