The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303029.1 | NP_001289958.1 | 13 - 14 | 97 | |
NM_007183.3 | NP_009114.1 | 12 - 13 | 97 | ||
GenBank mRNA | AF053719.1 | - | 12 - 13 | 97 | |
AK025067.1 | - | 12 - 13 | 97 | ||
AK223511.1 | - | 12 - 13 | 97 | ||
BC000081.2 | - | 12 - 13 | 2437 | 97 | |
FN421477.1 | - | 13 - 14 | 97 | ||
Z98265.1 | - | 12 - 13 | 2435 | 97 |