Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FECH
Entrez Gene ID
2235
Gene Name
ferrochelatase
Gene Aliases
EPP, FCE
Chromosome Location
Chr.18: 57544841 - 57586737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000140.3 NP_000131.2 10 - 11 1257 125
NM_001012515.2 NP_001012533.1 10 - 11 1275 125
XM_011525881.1 XP_011524183.1 9 - 10 1147 125
XM_011525882.2 XP_011524184.1 10 - 11 1240 125
XM_017025614.1 XP_016881103.1 9 - 10 1129 125
GenBank mRNA AK092416.1 - 10 - 11 1192 125
AK223190.1 - 10 - 11 1132 125
AK292937.1 - 10 - 11 1141 125
AK299643.1 - 10 - 11 1229 125
BC039841.1 - 10 - 11 1224 125
BT019958.1 - 10 - 11 1140 125
BT019959.1 - 10 - 11 1137 125
BX571744.1 - 10 - 11 1136 125
D00726.1 - 10 - 11 1169 125

View Full Product Details