The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002161.5 | NP_002152.2 | 22 - 23 | 2461 | 118 |
NM_013417.3 | NP_038203.2 | 22 - 23 | 2631 | 118 | |
NR_073446.1 | 21 - 22 | 2304 | 118 | ||
GenBank mRNA | AB209234.1 | - | 22 - 23 | 2370 | 118 |
AK293014.1 | - | 22 - 23 | 2388 | 118 | |
BC065552.1 | - | 21 - 22 | 2303 | 118 | |
BX537417.1 | - | 22 - 23 | 2399 | 118 | |
BX537429.1 | - | 22 - 23 | 2401 | 118 | |
BX538109.1 | - | 21 - 22 | 2244 | 118 | |
BX647071.1 | - | 22 - 23 | 2400 | 118 | |
BX647236.1 | - | 22 - 23 | 2393 | 118 | |
BX647430.1 | - | 22 - 23 | 2395 | 118 | |
BX648567.1 | - | 22 - 23 | 2401 | 118 | |
D28473.1 | - | 22 - 23 | 2549 | 118 |