Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
Chromosome Location
Chr.9: 92210207 - 92293756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002161.5 NP_002152.2 31 - 32 3559 146
NM_013417.3 NP_038203.2 31 - 32 3729 146
NR_073446.1 30 - 31 3402 146
GenBank mRNA AB209234.1 - 31 - 32 3468 146
AK293014.1 - 31 - 32 3486 146
AK301272.1 - 19 - 20 2039 146
AK308202.1 - 12 - 13 1296 146
BC008318.1 - 9 - 10 1064 146
BX537417.1 - 31 - 32 3497 146
BX537429.1 - 31 - 32 3499 146
BX647071.1 - 31 - 32 3498 146
BX647236.1 - 31 - 32 3491 146
BX647430.1 - 31 - 32 3493 146
BX648567.1 - 31 - 32 3499 146
D28473.1 - 31 - 32 3647 146
U04953.1 - 31 - 32 3661 146

View Full Product Details