The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002924.5 | NP_002915.3 | 16 - 17 | 1743 | 95 |
GenBank mRNA | BC022009.1 | - | 16 - 17 | 1696 | 95 |