The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033544.2 | NP_291022.2 | 2 - 3 | 162 | 68 |
GenBank mRNA | BC094739.1 | - | 2 - 3 | 154 | 68 |