The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308388.1 | NP_001295317.1 | 18 - 19 | 1537 | 77 |
NM_153635.2 | NP_705899.2 | 18 - 19 | 1518 | 77 | |
XM_011533386.2 | XP_011531688.1 | 17 - 18 | 1694 | 77 | |
XM_017005747.1 | XP_016861236.1 | 17 - 18 | 1278 | 77 | |
GenBank mRNA | BC035735.1 | - | 18 - 19 | 1518 | 77 |
BC130375.1 | - | 18 - 19 | 1389 | 77 | |
BU101942.1 | - | 3 - 4 | 77 |