The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308388.1 | NP_001295317.1 | 2 - 3 | 300 | 121 |
NM_153635.2 | NP_705899.2 | 2 - 3 | 281 | 121 | |
XM_011533386.2 | XP_011531688.1 | 1 - 2 | 457 | 121 | |
XM_011533388.2 | XP_011531690.1 | 1 - 2 | 457 | 121 | |
XM_011533389.2 | XP_011531691.1 | 1 - 2 | 457 | 121 | |
XM_017005747.1 | XP_016861236.1 | 2 - 3 | 103 | 121 | |
GenBank mRNA | BC035735.1 | - | 2 - 3 | 281 | 121 |
BC130375.1 | - | 2 - 3 | 152 | 121 | |
BM545574.1 | - | 2 - 3 | 281 | 121 | |
HY151604.1 | - | 2 - 3 | 301 | 121 |