The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003816.2 | NP_003807.1 | 3 - 4 | 325 | 146 |
NR_027638.1 | 3 - 4 | 325 | 146 | ||
NR_027639.1 | 3 - 4 | 325 | 146 | ||
NR_027878.1 | 3 - 4 | 325 | 146 | ||
XM_011544682.2 | XP_011542984.1 | 3 - 4 | 575 | 146 | |
XM_017013942.1 | XP_016869431.1 | 3 - 4 | 268 | 146 | |
GenBank mRNA | AF495383.1 | - | 3 - 4 | 280 | 146 |
BC126406.1 | - | 3 - 4 | 285 | 146 | |
BC143923.1 | - | 3 - 4 | 285 | 146 | |
BC143924.1 | - | 3 - 4 | 285 | 146 | |
U41766.1 | - | 3 - 4 | 325 | 146 |