Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYLPF
Entrez Gene ID
29895
Gene Name
myosin light chain, phosphorylatable, fast skeletal muscle
Gene Aliases
HUMMLC2B, MLC2B, MRLC2, MYL11
Chromosome Location
Chr.16: 30370934 - 30377991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324458.1 NP_001311387.1 4 - 5 317 66
NM_001324459.1 NP_001311388.1 4 - 5 345 66
NM_013292.4 NP_037424.2 2 - 3 180 66
GenBank mRNA AF363061.1 - 2 - 3 153 66
AK314082.1 - 2 - 3 180 66
BC012571.1 - 2 - 3 158 66
BI828249.1 - 4 - 5 346 66
BI837593.1 - 4 - 5 320 66
CR456963.1 - 1 - 2 99 66
M21812.1 - 2 - 3 160 66
X57543.1 - 2 - 3 160 66

View Full Product Details