The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242614.1 | NP_001229543.1 | 4 - 5 | 524 | 63 |
NM_031462.3 | NP_113650.2 | 4 - 5 | 512 | 63 | |
XM_017029890.1 | XP_016885379.1 | 4 - 5 | 376 | 63 | |
GenBank mRNA | AK125020.1 | - | 4 - 5 | 335 | 63 |
AK290200.1 | - | 4 - 5 | 288 | 63 | |
AK295217.1 | - | 4 - 5 | 330 | 63 | |
AL136580.1 | - | 4 - 5 | 402 | 63 | |
AY078165.1 | - | 4 - 5 | 320 | 63 | |
AY358837.1 | - | 4 - 5 | 385 | 63 | |
BC007569.1 | - | 3 - 4 | 172 | 63 | |
BC013029.2 | - | 3 - 4 | 172 | 63 | |
BC025729.1 | - | 5 - 6 | 1625 | 63 | |
BC030536.1 | - | 4 - 5 | 372 | 63 | |
DC323066.1 | - | 4 - 5 | 512 | 63 | |
KU178797.1 | - | 4 - 5 | 284 | 63 |