The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242614.1 | NP_001229543.1 | 6 - 7 | 669 | 105 |
NM_031462.3 | NP_113650.2 | 6 - 7 | 657 | 105 | |
NM_134445.3 | NP_604394.1 | 3 - 4 | 441 | 105 | |
NM_134446.3 | NP_604395.1 | 4 - 5 | 510 | 105 | |
XM_017029890.1 | XP_016885379.1 | 6 - 7 | 521 | 105 | |
XM_017029891.1 | XP_016885380.1 | 4 - 5 | 508 | 105 | |
GenBank mRNA | AK023776.1 | - | 5 - 6 | 105 | |
AK024278.1 | - | 2 - 3 | 164 | 105 | |
AK125020.1 | - | 6 - 7 | 480 | 105 | |
AK290200.1 | - | 6 - 7 | 433 | 105 | |
AK291375.1 | - | 4 - 5 | 323 | 105 | |
AL136580.1 | - | 6 - 7 | 547 | 104 | |
AY078165.1 | - | 6 - 7 | 465 | 105 | |
AY078166.1 | - | 4 - 5 | 318 | 105 | |
AY078167.1 | - | 3 - 4 | 249 | 105 | |
AY358837.1 | - | 6 - 7 | 530 | 105 | |
BC007569.1 | - | 5 - 6 | 317 | 105 | |
BC013029.2 | - | 5 - 6 | 317 | 105 | |
BC025729.1 | - | 7 - 8 | 1770 | 105 | |
BC030536.1 | - | 6 - 7 | 517 | 105 | |
KU178797.1 | - | 6 - 7 | 429 | 105 |