Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAP2
Entrez Gene ID
6891
Gene Name
transporter 2, ATP binding cassette subfamily B member
Gene Aliases
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11
Chromosome Location
Chr.6: 32821833 - 32838823 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000544.3 NP_000535.3 3 - 4 728 84
NM_001290043.1 NP_001276972.1 3 - 4 781 84
NM_018833.2 NP_061313.2 3 - 4 728 84
GenBank mRNA AB073779.1 - 2 - 3 606 84
AB208953.1 - 3 - 4 675 84
AF078671.1 - 3 - 4 685 84
AF105151.1 - 3 - 4 685 84
AK222823.1 - 2 - 3 637 84
AK223300.1 - 3 - 4 688 84
AK225657.1 - 3 - 4 709 84
AK225693.1 - 3 - 4 688 84
AK292963.1 - 3 - 4 711 84
BC002751.2 - 2 - 3 614 84
BT009906.1 - 2 - 3 606 84
FM246456.1 - 2 - 3 606 84
M74447.1 - 3 - 4 702 84
M84748.1 - 3 - 4 634 84
U07844.1 - 2 - 3 416 84
Z22935.1 - 3 - 4 634 84
Z22936.1 - 3 - 4 682 84

View Full Product Details