The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000544.3 | NP_000535.3 | 7 - 8 | 1392 | 124 |
NM_001290043.1 | NP_001276972.1 | 7 - 8 | 1445 | 124 | |
NM_018833.2 | NP_061313.2 | 7 - 8 | 1392 | 124 | |
GenBank mRNA | AB073779.1 | - | 6 - 7 | 1270 | 124 |
AB208953.1 | - | 7 - 8 | 1339 | 124 | |
AF078671.1 | - | 7 - 8 | 1349 | 124 | |
AF105151.1 | - | 7 - 8 | 1349 | 124 | |
AF176984.1 | - | 2 - 3 | 325 | 124 | |
AK222823.1 | - | 6 - 7 | 1301 | 124 | |
AK223300.1 | - | 7 - 8 | 1352 | 124 | |
AK225657.1 | - | 7 - 8 | 1373 | 124 | |
AK225693.1 | - | 7 - 8 | 1352 | 124 | |
AK292963.1 | - | 7 - 8 | 1375 | 124 | |
BC002751.2 | - | 6 - 7 | 1278 | 124 | |
BT009906.1 | - | 6 - 7 | 1270 | 124 | |
FM246456.1 | - | 6 - 7 | 1270 | 124 | |
M74447.1 | - | 7 - 8 | 1366 | 124 | |
M84748.1 | - | 7 - 8 | 1298 | 124 | |
U07844.1 | - | 6 - 7 | 1080 | 124 | |
Z22935.1 | - | 7 - 8 | 1298 | 124 | |
Z22936.1 | - | 7 - 8 | 1346 | 124 |