The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285439.1 | NP_001272368.1 | 10 - 11 | 919 | 111 |
NM_001285440.1 | NP_001272369.1 | 10 - 11 | 917 | 111 | |
NM_152756.4 | NP_689969.2 | 10 - 11 | 919 | 111 | |
XM_006714463.3 | XP_006714526.1 | 10 - 11 | 932 | 111 | |
XM_011514005.2 | XP_011512307.1 | 10 - 11 | 932 | 111 | |
XM_011514006.2 | XP_011512308.1 | 9 - 10 | 736 | 111 | |
XM_017009311.1 | XP_016864800.1 | 9 - 10 | 852 | 111 | |
XM_017009312.1 | XP_016864801.1 | 9 - 10 | 852 | 111 | |
XM_017009313.1 | XP_016864802.1 | 9 - 10 | 873 | 111 | |
GenBank mRNA | AK128358.1 | - | 9 - 10 | 852 | 111 |
AY515854.1 | - | 10 - 11 | 915 | 111 | |
BC137163.1 | - | 10 - 11 | 912 | 111 | |
BC137164.1 | - | 10 - 11 | 912 | 111 | |
BC144509.1 | - | 10 - 11 | 912 | 111 |