The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285439.1 | NP_001272368.1 | 3 - 4 | 224 | 117 |
NM_001285440.1 | NP_001272369.1 | 3 - 4 | 224 | 117 | |
NM_152756.4 | NP_689969.2 | 3 - 4 | 224 | 117 | |
XM_006714463.3 | XP_006714526.1 | 3 - 4 | 237 | 117 | |
XM_011514005.2 | XP_011512307.1 | 3 - 4 | 237 | 117 | |
XM_017009311.1 | XP_016864800.1 | 2 - 3 | 157 | 117 | |
XM_017009312.1 | XP_016864801.1 | 2 - 3 | 157 | 117 | |
XM_017009313.1 | XP_016864802.1 | 2 - 3 | 178 | 117 | |
XM_017009314.1 | XP_016864803.1 | 3 - 4 | 215 | 117 | |
XM_017009315.1 | XP_016864804.1 | 3 - 4 | 215 | 117 | |
GenBank mRNA | AY515854.1 | - | 3 - 4 | 220 | 117 |
BC029608.1 | - | 3 - 4 | 215 | 117 | |
BC137163.1 | - | 3 - 4 | 217 | 117 | |
BC137164.1 | - | 3 - 4 | 217 | 117 | |
BC144509.1 | - | 3 - 4 | 217 | 117 | |
CR749280.1 | - | 3 - 4 | 205 | 117 |