The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285439.1 | NP_001272368.1 | 12 - 13 | 1096 | 75 |
NM_001285440.1 | NP_001272369.1 | 12 - 13 | 1094 | 75 | |
NM_152756.4 | NP_689969.2 | 12 - 13 | 1096 | 75 | |
XM_006714463.3 | XP_006714526.1 | 12 - 13 | 1109 | 75 | |
XM_011514005.2 | XP_011512307.1 | 12 - 13 | 1109 | 75 | |
XM_011514006.2 | XP_011512308.1 | 11 - 12 | 913 | 75 | |
XM_017009311.1 | XP_016864800.1 | 11 - 12 | 1029 | 75 | |
XM_017009312.1 | XP_016864801.1 | 11 - 12 | 1029 | 75 | |
XM_017009313.1 | XP_016864802.1 | 11 - 12 | 1050 | 75 | |
XM_017009314.1 | XP_016864803.1 | 11 - 12 | 1019 | 75 | |
XM_017009315.1 | XP_016864804.1 | 11 - 12 | 1019 | 75 | |
GenBank mRNA | AY515854.1 | - | 12 - 13 | 1092 | 75 |
BC137163.1 | - | 12 - 13 | 1089 | 75 | |
BC137164.1 | - | 12 - 13 | 1089 | 75 | |
BC144509.1 | - | 12 - 13 | 1089 | 75 | |
CR749280.1 | - | 12 - 13 | 1075 | 75 |