The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040424.2 | NP_001035514.1 | 22 - 23 | 2950 | 59 |
NM_001282934.1 | NP_001269863.1 | 23 - 24 | 3010 | 59 | |
NM_022115.4 | NP_071398.3 | 29 - 30 | 3937 | 59 | |
NR_104257.1 | 23 - 24 | 2958 | 59 | ||
NR_104258.1 | 22 - 23 | 2838 | 59 | ||
NR_104260.1 | 22 - 23 | 2805 | 59 | ||
NR_135464.1 | 22 - 23 | 2946 | 59 | ||
XM_006724039.2 | XP_006724102.1 | 22 - 23 | 2792 | 59 | |
XM_006724040.2 | XP_006724103.1 | 21 - 22 | 2732 | 59 | |
XM_011529674.1 | XP_011527976.1 | 22 - 23 | 2822 | 59 | |
XM_011529675.1 | XP_011527977.1 | 23 - 24 | 2866 | 59 | |
XM_011529676.2 | XP_011527978.1 | 23 - 24 | 5275 | 59 | |
XM_011529677.2 | XP_011527979.1 | 23 - 24 | 2951 | 59 | |
XM_011529678.2 | XP_011527980.1 | 23 - 24 | 2995 | 59 | |
XM_011529679.2 | XP_011527981.1 | 23 - 24 | 3328 | 59 | |
XM_011529681.2 | XP_011527983.1 | 23 - 24 | 4232 | 59 | |
XM_011529683.1 | XP_011527985.1 | 28 - 29 | 3653 | 59 | |
XM_017028425.1 | XP_016883914.1 | 22 - 23 | 2805 | 59 | |
XM_017028426.1 | XP_016883915.1 | 22 - 23 | 5214 | 59 | |
GenBank mRNA | AB051812.1 | - | 22 - 23 | 2941 | 59 |
AB051813.1 | - | 23 - 24 | 3001 | 59 | |
AB051814.1 | - | 23 - 24 | 2949 | 59 | |
AB051815.1 | - | 22 - 23 | 2829 | 59 | |
AB209162.1 | - | 2 - 3 | 4885 | 59 | |
AF426259.1 | - | 2 - 3 | 398 | 59 | |
AF426260.1 | - | 2 - 3 | 398 | 59 | |
AY063456.1 | - | 4 - 5 | 421 | 59 | |
AY078498.2 | - | 29 - 30 | 3937 | 59 | |
BC067102.1 | - | 22 - 23 | 2805 | 59 | |
CA949877.1 | - | 2 - 3 | 107 | 59 | |
HY146343.1 | - | 3 - 4 | 333 | 59 |