The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040424.2 | NP_001035514.1 | 12 - 13 | 1759 | 74 |
NM_001282934.1 | NP_001269863.1 | 13 - 14 | 1819 | 74 | |
NM_022115.4 | NP_071398.3 | 19 - 20 | 2746 | 74 | |
NR_104257.1 | 13 - 14 | 1767 | 74 | ||
NR_104258.1 | 12 - 13 | 1647 | 74 | ||
NR_104260.1 | 12 - 13 | 1614 | 74 | ||
NR_135464.1 | 12 - 13 | 1755 | 74 | ||
XM_006724039.2 | XP_006724102.1 | 13 - 14 | 1733 | 74 | |
XM_006724040.2 | XP_006724103.1 | 12 - 13 | 1673 | 74 | |
XM_011529674.1 | XP_011527976.1 | 12 - 13 | 1631 | 74 | |
XM_011529675.1 | XP_011527977.1 | 13 - 14 | 1675 | 74 | |
XM_011529676.2 | XP_011527978.1 | 13 - 14 | 4084 | 74 | |
XM_011529677.2 | XP_011527979.1 | 13 - 14 | 1760 | 74 | |
XM_011529678.2 | XP_011527980.1 | 13 - 14 | 1804 | 74 | |
XM_011529679.2 | XP_011527981.1 | 13 - 14 | 2137 | 74 | |
XM_011529681.2 | XP_011527983.1 | 13 - 14 | 3041 | 74 | |
XM_011529683.1 | XP_011527985.1 | 18 - 19 | 2462 | 74 | |
XM_017028425.1 | XP_016883914.1 | 12 - 13 | 1614 | 74 | |
XM_017028426.1 | XP_016883915.1 | 12 - 13 | 4023 | 74 | |
GenBank mRNA | AB051812.1 | - | 12 - 13 | 1750 | 74 |
AB051813.1 | - | 13 - 14 | 1810 | 74 | |
AB051814.1 | - | 13 - 14 | 1758 | 74 | |
AB051815.1 | - | 12 - 13 | 1638 | 74 | |
AF276513.1 | - | 19 - 20 | 2746 | 74 | |
AY078498.2 | - | 19 - 20 | 2746 | 74 | |
BC067102.1 | - | 12 - 13 | 1614 | 74 | |
CF137278.1 | - | 1 - 2 | 133 | 74 |