The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040424.2 | NP_001035514.1 | 1 - 2 | 214 | 65 |
NM_001282934.1 | NP_001269863.1 | 1 - 2 | 214 | 65 | |
NR_104257.1 | 1 - 2 | 214 | 65 | ||
NR_104258.1 | 1 - 2 | 214 | 65 | ||
XM_006724039.2 | XP_006724102.1 | 1 - 2 | 128 | 65 | |
XM_006724040.2 | XP_006724103.1 | 1 - 2 | 128 | 65 | |
XM_011529674.1 | XP_011527976.1 | 1 - 2 | 128 | 65 | |
GenBank mRNA | AB051812.1 | - | 1 - 2 | 205 | 65 |
AB051813.1 | - | 1 - 2 | 205 | 65 | |
AB051814.1 | - | 1 - 2 | 205 | 65 | |
AB051815.1 | - | 1 - 2 | 205 | 65 |