The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040147.2 | NP_001035237.1 | 4 - 5 | 424 | 115 |
NM_001261830.1 | NP_001248759.1 | 4 - 5 | 399 | 115 | |
NM_001261831.1 | NP_001248760.1 | 3 - 4 | 641 | 115 | |
NM_003784.3 | NP_003775.1 | 4 - 5 | 692 | 115 | |
GenBank mRNA | AF027866.1 | - | 4 - 5 | 696 | 115 |
AK290673.1 | - | 4 - 5 | 396 | 115 | |
AK290876.1 | - | 4 - 5 | 643 | 115 | |
AK300828.1 | - | 3 - 4 | 595 | 115 | |
BC069417.1 | - | 4 - 5 | 644 | 115 | |
BC069442.1 | - | 4 - 5 | 644 | 115 | |
BC069547.1 | - | 4 - 5 | 644 | 115 | |
BC106743.2 | - | 4 - 5 | 552 | 115 | |
BC106744.2 | - | 4 - 5 | 552 | 115 | |
CD702645.1 | - | 4 - 5 | 454 | 115 | |
D88575.1 | - | 4 - 5 | 405 | 115 | |
DA577978.1 | - | 4 - 5 | 400 | 115 |