Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NR6A1
- Entrez Gene ID
-
2649
- Gene Name
- nuclear receptor subfamily 6 group A member 1
- Gene Aliases
-
CT150, GCNF, GCNF1, NR61, RTR, hGCNF, hRTR
- Chromosome Location
-
Chr.9: 124517275 - 124771310 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF004291.1 |
- |
1 - 2 |
207 |
67 |
View Full Product Details