The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127443.1 | NP_001120915.1 | 1 - 2 | 33 | 92 |
GenBank mRNA | DA566475.1 | - | 1 - 2 | 33 | 92 |
L06850.1 | - | 1 - 2 | 45 | 92 |