The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166698.1 | NP_001160170.1 | 1 - 2 | 190 | 79 |
XM_011539491.2 | XP_011537793.1 | 1 - 2 | 118 | 79 | |
GenBank mRNA | AK316542.1 | - | 1 - 2 | 186 | 79 |