The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031732.2 | NP_001026902.1 | 2 - 3 | 475 | 123 |
NM_133370.2 | NP_588611.2 | 2 - 3 | 475 | 123 | |
XM_005265706.2 | XP_005265763.1 | 2 - 3 | 475 | 123 | |
XM_005265708.2 | XP_005265765.1 | 2 - 3 | 475 | 123 | |
GenBank mRNA | AK056430.1 | - | 2 - 3 | 460 | 123 |
BC041119.1 | - | 2 - 3 | 405 | 123 | |
BC053863.1 | - | 2 - 3 | 140 | 123 | |
DA191646.1 | - | 2 - 3 | 475 | 123 |