The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_148910.2 | NP_683708.1 | 2 - 3 | 231 | 80 |
XM_017017162.1 | XP_016872651.1 | 2 - 3 | 252 | 80 | |
GenBank mRNA | AF378129.1 | - | 2 - 3 | 212 | 80 |
AF410783.1 | - | 2 - 3 | 208 | 80 |