The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011544015.2 | XP_011542317.1 | 3 - 4 | 1950 | 61 |
XM_016999986.1 | XP_016855475.1 | 2 - 3 | 2672 | 61 |