Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GUSBP10
Entrez Gene ID
642006
Gene Name
glucuronidase, beta pseudogene 10
Gene Aliases
-
Chromosome Location
Chr.7: 57165670 - 57180156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_030766.1 1 - 2 85
GenBank mRNA EG328188.1 - 1 - 2 85
EG328189.1 - 1 - 2 124 85

View Full Product Details